2,3,4,6-Tetrafluoroaniline 95%

Supplier: Ambeed
Danger

A825508-1G A825508-5G
77569-536EA 66.95 USD
77569-536 77618-314
2,3,4,6-Tetrafluoroaniline 95%
2,3,4,6-Tetrafluoroaniline

Precaución: For research use only.

Formula: C₆H₃F₄N
MW: 165.09 g/mol
MDL Number: MFCD00069077
CAS Number: 363-73-5
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR