Menthyl acetate 98%

Supplier: Ambeed
Danger

Synonyms: (±)-Menthol acetate, (±)-Menthyl acetate, rel-(1R,2S,5R)-2-Isopropyl-5-methylcyclohexyl acetate, dl-Menthyl acetate

A819714-25G A819714-100G
77569-528EA 21 USD
77569-528 77880-620
Menthyl acetate 98%
Menthyl acetate

Precaución: For research use only.

Formula: C₁₂H₂₂O₂
MDL Number: MFCD00062975
CAS Number: 89-48-5
UN: 3082
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR