tert-Butyl 6-hydroxy-1,4-oxazepane-4-carboxylate 98%

Supplier: Ambeed
Warning

A808722-1G A808722-5G A808722-250MG
77569-510EA 82.4 USD
77569-510 77573-582 77684-762
tert-Butyl 6-hydroxy-1,4-oxazepane-4-carboxylate 98%
tert-Butyl 6-hydroxy-1,4-oxazepane-4-carboxylate

Precaución: For research use only.

Formula: C₁₀H₁₉NO₄
MW: 217.26 g/mol
MDL Number: MFCD18711407
CAS Number: 1272758-40-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR