tert-Butyl 2-(3-(2-aminoethoxy)propoxy)acetate 98%

Supplier: Ambeed
Warning

Synonyms: NH2-PEG2-CH2-Boc

A802211-250MG A802211-1G A802211-100MG
77569-504EA 374 USD
77569-504 77622-956 77784-088
tert-Butyl 2-(3-(2-aminoethoxy)propoxy)acetate 98%
tert-Butyl 2-(3-(2-aminoethoxy)propoxy)acetate

Precaución: For research use only.

Formula: C₁₁H₂₃NO₄
MDL Number: MFCD30741911
CAS Number: 1948273-09-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR