Methyl 3-bromo-1H-pyrazolo[4,3-c]pyridine-6-carboxylate 96%

Supplier: Ambeed
Warning

A801532-1G A801532-100MG
77569-502EA 629.33 USD
77569-502 77674-168
Methyl 3-bromo-1H-pyrazolo[4,3-c]pyridine-6-carboxylate 96%
Methyl 3-bromo-1H-pyrazolo[4,3-c]pyridine-6-carboxylate

Precaución: For research use only.

Formula: C₈H₆BrN₃O₂
MW: 256.056 g/mol
MDL Number: MFCD20040381
CAS Number: 1206979-28-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR