Fmoc-4-(boc-methoxy)-L-phenylalanine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
N-Fmoc-4-(tert-butoxycarbonylmethoxy)-L-phenylalanine, (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(4-(2-(tert-butoxy)-2-oxoethoxy)phenyl)propanoic acid
A801502-1G
A801502-5G
77569-500EA
89.61
USD
77569-500
77670-132
Fmoc-4-(boc-methoxy)-L-phenylalanine 98%
Fmoc-4-(boc-methoxy)-L-phenylalanine
Precaución: For research use only.
Formula:
C₃₀H₃₁NO₇ MW: 517.58 g/mol |
MDL Number:
MFCD07781259 CAS Number: 181951-92-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...