5-(Bromomethyl)benzo[d]oxazole 97%

Supplier: Ambeed
Danger

A793817-1G A793817-250MG
77569-486EA 349 USD
77569-486 77573-542
5-(Bromomethyl)benzo[d]oxazole 97%
5-(Bromomethyl)benzo[d]oxazole

Precaución: For research use only.

Formula: C₈H₆BrNO
MDL Number: MFCD16036446
CAS Number: 181038-98-2
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR