Phenylcarbamic chloride 95%

Supplier: Ambeed
Danger

A785986-1G A785986-100MG A785986-250MG
77569-480EA 532 USD
77569-480 77884-726 77884-727
Phenylcarbamic chloride 95%

Precaución: For research use only.

Formula: C₇H₆ClNO
MDL Number: MFCD24368249
CAS Number: 2040-76-8
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR