6-Bromo-1H-pyrido[2,3-b][1,4]oxazin-2(3H)-one 98%

Supplier: Ambeed
Warning

A776240-1G A776240-250MG
77569-470EA 236.9 USD
77569-470 77684-716
6-Bromo-1H-pyrido[2,3-b][1,4]oxazin-2(3H)-one 98%
6-Bromo-1H-pyrido[2,3-b][1,4]oxazin-2(3H)-one

Precaución: For research use only.

Formula: C₇H₅BrN₂O₂
MW: 229.031 g/mol
MDL Number: MFCD15526939
CAS Number: 1245708-13-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR