N-(3-(5-Bromo-1H-pyrrolo[2,3-b]pyridine-3-carbonyl)-2,4-difluorophenyl)propane-1-sulfonamide 98%

Supplier: Ambeed
Warning

Synonyms: B-Raf IN 11

A768926-1G A768926-100MG A768926-250MG A768926-5G
77569-458EA 120.51 USD
77569-458 77684-707 77684-708 77684-709
N-(3-(5-Bromo-1H-pyrrolo[2,3-b]pyridine-3-carbonyl)-2,4-difluorophenyl)propane-1-sulfonamide 98%
N-(3-(5-Bromo-1H-pyrrolo[2,3-b]pyridine-3-carbonyl)-2,4-difluorophenyl)propane-1-sulfonamide

Precaución: For research use only.

Formula: C₁₇H₁₄BrF₂N₃O₃S
MW: 458.27 g/mol
MDL Number: MFCD18207768
CAS Number: 918504-27-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR