19-Bromo-2,5,8,11,14,17-hexaoxanonadecane 95%

Supplier: Ambeed
Warning

A755391-5G A755391-1G A755391-250MG
77569-442EA 422 USD
77569-442 77598-980 77622-798
19-Bromo-2,5,8,11,14,17-hexaoxanonadecane 95%
19-Bromo-2,5,8,11,14,17-hexaoxanonadecane

Precaución: For research use only.

Formula: C₁₃H₂₇BrO₆
MDL Number: MFCD22683286
CAS Number: 125562-29-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR