Propargyl-PEG3-Bromide 95%

Supplier: Ambeed
Warning

Synonyms: 3-(2-(2-(2-Bromoethoxy)ethoxy)ethoxy)prop-1-yne

A754168-1G A754168-250MG A754168-5G
77569-438EA 182 USD
77569-438 77598-934 77598-936
Propargyl-PEG3-Bromide 95%
Propargyl-PEG3-Bromide

Precaución: For research use only.

Formula: C₉H₁₅BrO₃
MDL Number: MFCD28505517
CAS Number: 203740-63-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR