Biotin-PEG3-NHS ester 95%

Supplier: Ambeed
Warning

Synonyms: 2,5-Dioxopyrrolidin-1-yl 14-oxo-18-((3aS,4S,6aR)-2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)-4,7,10-trioxa-13-azaoctadecanoate

A753922-250MG A753922-100MG A753922-1G A753922-5G
77569-436EA 125 USD
77569-436 77622-734 77622-736 77783-903
Biotin-PEG3-NHS ester 95%
Biotin-PEG3-NHS ester

Precaución: For research use only.

Formula: C₂₃H₃₆N₄O₉S
MDL Number: MFCD11974977
CAS Number: 1253286-56-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR