5-(Trifluoromethyl)-1H-imidazole-2-carboxylic acid 95%

Supplier: Ambeed
Warning

A751900-250MG A751900-100MG
77569-428EA 120 USD
77569-428 77893-485
5-(Trifluoromethyl)-1H-imidazole-2-carboxylic acid 95%
5-(Trifluoromethyl)-1H-imidazole-2-carboxylic acid

Precaución: For research use only.

Formula: C₅H₃F₃N₂O₂
MDL Number: MFCD14583619
CAS Number: 915230-46-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR