tert-Butyl 1-bromo-3,6,9,12,15-pentaoxaoctadecan-18-oate 96%
Supplier: Ambeed
|
Warning
|
A751446-5G
A751446-1G
A751446-250MG
77569-426EA
259
USD
77569-426
77598-900
77622-686
tert-Butyl 1-bromo-3,6,9,12,15-pentaoxaoctadecan-18-oate 96%
tert-Butyl 1-bromo-3,6,9,12,15-pentaoxaoctadecan-18-oate
Precaución: For research use only.
Formula:
C₁₇H₃₃BrO₇ |
MDL Number:
MFCD22683304 CAS Number: 1415800-38-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...