2-(3-Chloropropyl)-1,3-dioxolane 98%

Supplier: Ambeed
Warning

Synonyms: 2-(3-chloropropyl)-1,3-dioxolane, 4-Chlorobutyraldehyde ethylene Acetal

A750681-25G A750681-5G
77569-422EA 213.21 USD
77569-422 77747-480
2-(3-Chloropropyl)-1,3-dioxolane 98%
2-(3-Chloropropyl)-1,3-dioxolane

Precaución: For research use only.

Formula: C₆H₁₁ClO₂
MW: 150.6 g/mol
MDL Number: MFCD00043135
CAS Number: 16686-11-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR