Methyl phenyl sulphoxide 98%

Supplier: Ambeed
Danger

Synonyms: (Methylsulphinyl)benzene

A737495-25G A737495-1G A737495-5G
77569-392EA 54 USD
77569-392 77882-620 77882-621
Methyl phenyl sulphoxide 98%
Methyl phenyl sulphoxide

Precaución: For research use only.

Formula: C₇H₈OS
MDL Number: MFCD00002088
CAS Number: 1193-82-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR