4-(1,3-Dioxolan-2-yl)benzonitrile 97%

Supplier: Ambeed
Danger

A735561-1G A735561-5G
77569-388EA 56 USD
77569-388 77569-390
4-(1,3-Dioxolan-2-yl)benzonitrile 97%
4-(1,3-Dioxolan-2-yl)benzonitrile

Precaución: For research use only.

Formula: C₁₀H₉NO₂
MDL Number: MFCD00052570
CAS Number: 66739-89-7
UN: 3439
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR