3-(Trimethylsilyl)-2-naphthyl trifluoromethanesulfonate 95%

Supplier: Ambeed
Danger

A721772-1G A721772-100MG A721772-250MG A721772-5G
77569-370EA 129 USD
77569-370 77900-794 77901-699 77901-732
3-(Trimethylsilyl)-2-naphthyl trifluoromethanesulfonate 95%
3-(Trimethylsilyl)-2-naphthyl trifluoromethanesulfonate

Precaución: For research use only.

Formula: C₁₄H₁₅F₃O₃SSi
MDL Number: MFCD10566935
CAS Number: 780820-43-1
UN: 3265
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR