Ethyl-2,3-butadienoate 97%

Supplier: Ambeed
Danger

Synonyms: Ethyl buta-2,3-dienoate, 2,3-Butadienoic Acid Ethyl Ester

A714405-1G A714405-5G
77569-366EA 54.59 USD
77569-366 77684-633
Ethyl-2,3-butadienoate 97%
Ethyl-2,3-butadienoate

Precaución: For research use only.

Formula: C₆H₈O₂
MW: 112.13 g/mol
MDL Number: MFCD01863567
CAS Number: 14369-81-4
UN: 3272
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR