Piperonyl alcohol 98%

Supplier: Ambeed
Warning

A712929-100G A712929-5G A712929-25G A712929-500G A712929-10G
77569-364EA 54 USD
77569-364 77573-314 77602-814 77602-816 77669-916
Piperonyl alcohol 98%
Piperonyl alcohol

Precaución: For research use only.

Formula: C₈H₈O₃
MDL Number: MFCD00005836
CAS Number: 495-76-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR