Fmoc-D-Pentafluorophenylalanine 98%

Supplier: Ambeed
Warning

Synonyms: Fmoc-D-Phe(F5)-OH, Fmoc-D-Phe(F)₅-OH

A704353-5G A704353-1G
77569-342EA 221 USD
77569-342 77882-563
Fmoc-D-Pentafluorophenylalanine 98%
Fmoc-D-Pentafluorophenylalanine

Precaución: For research use only.

Formula: C₂₄H₁₆F₅NO₄
MDL Number: MFCD00672558
CAS Number: 198545-85-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR