2-Ethylhexyl-3-mercaptopropionate 98%

Supplier: Ambeed
Danger

A701196-5G A701196-25G
77569-334EA 7 USD
77569-334 77573-266
2-Ethylhexyl-3-mercaptopropionate 98%
2-Ethylhexyl-3-mercaptopropionate

Precaución: For research use only.

Formula: C₁₁H₂₂O₂S
MDL Number: MFCD00087933
CAS Number: 50448-95-8
UN: 3082
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR