(Ethoxycarbonylmethyl)dimethylsulfonium bromide 98%

Supplier: Ambeed
Danger

Synonyms: (2-Ethoxy-2-oxoethyl)(dimethyl)sulphonium bromide

A694968-1G A694968-5G
77569-322EA 6 USD
77569-322 77747-399
(Ethoxycarbonylmethyl)dimethylsulfonium bromide 98%
(Ethoxycarbonylmethyl)dimethylsulfonium bromide

Precaución: For research use only.

Formula: CH₃CH₂O₂CCH₂S(CH₃)₂Br
MDL Number: MFCD00830557
CAS Number: 5187-82-6
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR