Mal-amido-PEG8-acid 95%

Supplier: Ambeed
Warning

Synonyms: Mal-amido-PEG8-C2-acid, 1-(2,5-Dioxo-2,5-dihydro-1H-pyrrol-1-yl)-3-oxo-7,10,13,16,19,22,25,28-octaoxa-4-azahentriacontan-31-oic acid

A680616-250MG A680616-1G A680616-100MG
77569-298EA 69.01 USD
77569-298 77614-102 77684-582
Mal-amido-PEG8-acid 95%
Mal-amido-PEG8-acid

Precaución: For research use only.

Formula: C₂₆H₄₄N₂O₁₃
MW: 592.63 g/mol
MDL Number: MFCD13184956
CAS Number: 1334177-86-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR