(±)-trans-1,2-Cyclopentanediol 98%

Supplier: Ambeed
Warning

A679837-5G A679837-1G
77569-290EA 46 USD
77569-290 77573-200
(±)-trans-1,2-Cyclopentanediol 98%
(±)-trans-1,2-Cyclopentanediol

Precaución: For research use only.

Formula: C₅H₁₀O₂
MDL Number: MFCD00082582
CAS Number: 5057-99-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR