Potassium 4-fluorobenzyl-trifluoroborate 95%

Supplier: Ambeed
Warning

A673256-1G A673256-250MG
77569-276EA 45 USD
77569-276 77902-049
Potassium 4-fluorobenzyl-trifluoroborate 95%
Potassium 4-fluorobenzyl-trifluoroborate

Precaución: For research use only.

Formula: C₇H₆BF₄·K
MDL Number: MFCD04112715
CAS Number: 1494466-28-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR