Benzo[b]thiophene-4-carbaldehyde 97%

Supplier: Ambeed
Warning

A657969-1G A657969-250MG A657969-100MG
77569-248EA 184.37 USD
77569-248 77673-788 77673-786
Benzo[b]thiophene-4-carbaldehyde 97%
Benzo[b]thiophene-4-carbaldehyde

Precaución: For research use only.

Formula: C₉H₆OS
MW: 162.20 g/mol
MDL Number: MFCD18432490
CAS Number: 10133-25-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR