2-Bromo-4-iodobenzaldehyde 97%

Supplier: Ambeed
Warning

A655574-5G A655574-1G
77569-242EA 124.63 USD
77569-242 77686-620
2-Bromo-4-iodobenzaldehyde 97%
2-Bromo-4-iodobenzaldehyde

Precaución: For research use only.

Formula: C₇H₄BrIO
MW: 310.9200 g/mol
MDL Number: MFCD07779025
CAS Number: 261903-03-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR