cis-1,2-Bis(phenylsulfonyl)ethylene 95%

Supplier: Ambeed
Warning

Synonyms: (Z)-1,2-Bis(phenylsulphonyl)ethene

A655517-5G A655517-25G A655517-1G
77569-240EA 122 USD
77569-240 77893-316 77893-315
cis-1,2-Bis(phenylsulfonyl)ethylene 95%
cis-1,2-Bis(phenylsulfonyl)ethylene

Precaución: For research use only.

Formula: C₁₄H₁₂O₄S₂
MDL Number: MFCD00010140
CAS Number: 963-15-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR