1-Bromodibenzothiophene 97%

Supplier: Ambeed
Warning

A645395-5G A645395-1G
77569-232EA 266 USD
77569-232 77747-328
1-Bromodibenzothiophene 97%
1-Bromodibenzothiophene

Precaución: For research use only.

Formula: C₁₂H₇BrS
MDL Number: MFCD18451629
CAS Number: 65642-94-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR