2-Mercapto-N-methylbenzamide 98%

Supplier: Ambeed
Danger

A638243-5G A638243-1G A638243-25G
77569-222EA 20 USD
77569-222 77594-958 77673-732
2-Mercapto-N-methylbenzamide 98%
2-Mercapto-N-methylbenzamide

Precaución: For research use only.

Formula: C₈H₉NOS
MDL Number: MFCD00136334
CAS Number: 20054-45-9
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR