3,5-Dichlorobenzylamine 98%

Supplier: Ambeed
Danger

A632308-1G A632308-5G
77569-204EA 9 USD
77569-204 77686-578
3,5-Dichlorobenzylamine 98%
3,5-Dichlorobenzylamine

Precaución: For research use only.

Formula: C₇H₇Cl₂N
MDL Number: MFCD00052681
CAS Number: 39989-43-0
UN: 2735
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR