1,3-Dioxan-2-one 98%

Supplier: Ambeed
Warning

A621661-25G A621661-1G A621661-100G A621661-500G A621661-5G
77569-192EA 86 USD
77569-192 77573-018 77886-499 77886-500 77886-501
1,3-Dioxan-2-one 98%
1,3-Dioxan-2-one

Precaución: For research use only.

Formula: C₄H₆O₃
MDL Number: MFCD00010440
CAS Number: 2453-03-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR