4-Bromo-1-tritylimidazole 95%

Supplier: Ambeed
Warning

Synonyms: 4-Bromo-1-trityl-1H-imidazole

A619808-1G A619808-5G
77569-184EA 57.68 USD
77569-184 77569-186
4-Bromo-1-tritylimidazole 95%
4-Bromo-1-tritylimidazole

Precaución: For research use only.

Formula: C₂₂H₁₇BrN₂
MW: 389.2878 g/mol
MDL Number: MFCD00185657
CAS Number: 87941-55-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR