2-Bromo-5-(trifluoromethyl)-1,3,4-thiadiazole 98%

Supplier: Ambeed
Danger

A619703-1G A619703-250MG
77569-182EA 93.73 USD
77569-182 77669-664
2-Bromo-5-(trifluoromethyl)-1,3,4-thiadiazole 98%
2-Bromo-5-(trifluoromethyl)-1,3,4-thiadiazole

Precaución: For research use only.

Formula: C₃BrF₃N₂S
MDL Number: MFCD00015535
CAS Number: 37461-61-3
UN: 1993
ADR: 3,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR