2-[(Prop-2-ynyloxy)methyl]oxirane 92%

Supplier: Ambeed
Danger

Synonyms: 2,3-Epoxypropyl Propargyl ether, Glycidyl Propargyl ether, Glycidyl 2-Propynyl ether

A618069-25G A618069-5G A618069-1G
77569-174EA 137 USD
77569-174 77602-560 77880-321
2-[(Prop-2-ynyloxy)methyl]oxirane 92%
2-[(Prop-2-ynyloxy)methyl]oxirane

Precaución: For research use only.

Formula: C₆H₈O₂
MW: 112.13 g/mol
MDL Number: MFCD00099300
CAS Number: 18180-30-8
UN: 2810
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR