Fmoc-D-Lys(Dde)-OH 98%

Supplier: Ambeed
Warning

A617528-1G A617528-5G
77569-170EA 21.63 USD
77569-170 77569-172
Fmoc-D-Lys(Dde)-OH 98%
Fmoc-D-Lys(Dde)-OH

Precaución: For research use only.

Formula: C₃₁H₃₆N₂O₆
MW: 532.6 g/mol
MDL Number: MFCD01862894
CAS Number: 333973-51-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR