5-Azaspiro[2.4]heptane hydrochloride 98%

Supplier: Ambeed
Warning

A616791-250MG A616791-1G
77569-166EA 100.94 USD
77569-166 77613-920
5-Azaspiro[2.4]heptane hydrochloride 98%
5-Azaspiro[2.4]heptane hydrochloride

Precaución: For research use only.

Formula: C₆H₁₂ClN
MW: 133.62 g/mol
MDL Number: MFCD21099571
CAS Number: 3659-21-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR