2-Bromobutyryl bromide 98%

Supplier: Ambeed
Danger

A613707-25G A613707-5G A613707-100G
77569-164EA 158 USD
77569-164 77747-268 77747-267
2-Bromobutyryl bromide 98%
2-Bromobutyryl bromide

Precaución: For research use only.

Formula: C₄H₆Br₂O
MDL Number: MFCD00000154
CAS Number: 26074-52-2
UN: 3265
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR