(3-Methoxypyridin-2-yl)boronic acid 95+%

Supplier: Ambeed
Warning

A605668-100MG A605668-1G A605668-250MG
77569-154EA 138 USD
77569-154 77572-958 77572-960
(3-Methoxypyridin-2-yl)boronic acid 95+%
(3-Methoxypyridin-2-yl)boronic acid

Precaución: For research use only.

Formula: C₆H₈BNO₃
MDL Number: MFCD07367559
CAS Number: 500707-34-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR