2,3-Dimercapto-1-propanesulfonic acid sodium salt monohydrate 97%

Supplier: Ambeed
Warning

Synonyms: DL-2,3-Dimercapto-1-propanesulphonic acid, sodium salt monohydrate, DMPS, Sodium 2,3-dimercaptopropane-1-sulphonate hydrate, 2,3-Dimercaptopropanesulphonic acid sodium salt, Sodium 2,3-Dimercaptopropanesulphonate monohydrate

A599319-5G A599319-1G
77569-142EA 178 USD
77569-142 77594-824
2,3-Dimercapto-1-propanesulfonic acid sodium salt monohydrate 97%
2,3-Dimercapto-1-propanesulfonic acids odium salt monohydrate

Precaución: For research use only.

Formula: C₃H₉NaO₄S₃·H₂O
MDL Number: MFCD00149543
CAS Number: 207233-91-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR