Cobalt(II) 1,2,3,4,8,9,10,11,15,16,17,18,22,23,24,25-hexadecafluoro-29H,31H-phthalocyanine 98%

Supplier: Ambeed
Warning

A597462-100MG A597462-1G
77569-138EA 111 USD
77569-138 77749-232
Cobalt(II) 1,2,3,4,8,9,10,11,15,16,17,18,22,23,24,25-hexadecafluoro-29H,31H-phthalocyanine 98%
Cobalt(II) 1,2,3,4,8,9,10,11,15,16,17,18,22,23,24,25-hexadecafluoro-29H,31H-phthalocyanine

Precaución: For research use only.

Formula: C₃₂CoF₁₆N₈
MW: 859.30 g/mol
MDL Number: MFCD00274641
CAS Number: 52629-20-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR