Rose Bengal BR

Supplier: Ambeed
Warning

A596993-5G A596993-25G A596993-100G
77569-136EA 15 USD
77569-136 77569-134 77669-576
Rose Bengal BR
Rose Bengal

Precaución: For research use only.

Formula: C₂₀H₂Cl₄I₄O₅Na₂
MDL Number: MFCD00151169
CAS Number: 632-69-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR