2,5-Dichloro-p-benzoquinone 97%

Supplier: Ambeed
Warning

A593551-10G A593551-1G A593551-25G A593551-5G
77569-130EA 110 USD
77569-130 77900-918 77900-697 77900-971
2,5-Dichloro-p-benzoquinone 97%
2,5-Dichloro-p-benzoquinone

Precaución: For research use only.

Formula: C₆H₂Cl₂O₂
MDL Number: MFCD00041736
CAS Number: 615-93-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR