3-(Hydroxymethyl)pyrrolidin-2-one 98%

Supplier: Ambeed
Warning

A589918-1G A589918-100MG A589918-250MG A589918-5G
77569-118EA 270 USD
77569-118 77891-168 77891-169 77891-170
3-(Hydroxymethyl)pyrrolidin-2-one 98%
3-(Hydroxymethyl)pyrrolidin-2-one

Precaución: For research use only.

Formula: C₅H₉NO₂
MDL Number: MFCD18074198
CAS Number: 76220-94-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR