Mal-PEG2-NHS ester 95%

Supplier: Ambeed
Warning

A584523-100MG A584523-1G A584523-250MG A584523-5G
77569-114EA 59 USD
77569-114 77569-116 77613-808 77900-994
Mal-PEG2-NHS ester 95%
Mal-PEG2-NHS ester

Precaución: For research use only.

Formula: C₁₅H₁₈N₂O₈
MDL Number: MFCD24539465
CAS Number: 1433997-01-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR