N-Methyl-M-anisidine 98%

Supplier: Ambeed
Warning

Synonyms: 3-(Methylamino)anisole, N-Methyl-3-methoxyaniline, 3-Methoxy-N-methylaniline, N-Methyl-3-anisidine

A579742-1G A579742-5G
77569-106EA 28.84 USD
77569-106 77749-193
N-Methyl-M-anisidine 98%
N-Methyl-M-anisidine

Precaución: For research use only.

Formula: C₈H₁₁NO
MW: 137.18 g/mol
MDL Number: MFCD05665953
CAS Number: 14318-66-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR