Methyl 5-(chlorosulfonyl)picolinate 95%

Supplier: Ambeed
Danger

A567517-1G A567517-250MG
77569-090EA 640.66 USD
77569-090 77684-423
Methyl 5-(chlorosulfonyl)picolinate 95%
Methyl 5-(chlorosulfonyl)picolinate

Precaución: For research use only.

Formula: C₇H₆ClNO₄S
MW: 235.645 g/mol
MDL Number: MFCD18254200
CAS Number: 1063733-25-4
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR