Nitro blue tetrazolium chloride (NBT) ≥98% (by HPLC) (contains <7% EtOH)

Supplier: Ambeed
Danger

Synonyms: 5,5'-Diphenyl-3,3'-bis(4-nitrophenyl)-2,2'-(3,3'-dimethoxybiphenyl-4,4'-ylene)ditetrazolium dichloride

A543531-1G A543531-25G
77569-060EA 45 USD
77569-060 77673-454
Nitro blue tetrazolium chloride (NBT) ≥98% (by HPLC) (contains <7% EtOH)
Nitro blue tetrazolium chloride (NBT)

Precaución: For research use only.

Formula: C₄₀H₃₀Cl₂N₁₀O₆
MW: 817.65 g/mol
Melting Pt: 200
MDL Number: MFCD00012159
CAS Number: 298-83-9
UN: 1759
ADR: 8 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR